Learn More
Medchemexpress LLC Kansuinine A | 57701-86-7 | 99.8% | 730.75 | 1 MG
SDP
Catalog No. 5000438034
Encompass_Preferred
Kansuinine A inhibits IL-6-induced Stat3 activation. It possesses antiviral and anticancer activity.
  • Inhibits IL-6-induced Stat3 activation
  • Possesses antiviral activity
  • Possesses anticancer activity
Catalog No. 50-004-38034 Supplier Medchemexpress LLC Supplier No. HY1264211MG
Kansuinine A inhibits IL-6-induced Stat3 activation. It possesses antiviral and anticancer activity.
  • Inhibits IL-6-induced Stat3 activation
  • Possesses antiviral activity
  • Possesses anticancer activity