Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Kansuinine A | 57701-86-7 | 99.8% | 730.75 g·mol⁻1 | C37H46O15 | 5 MG
SDP

Catalog No. 5000264465
Encompass_Preferred

Kansuinine A is a natural diterpenoid research compound that inhibits IL-6-induced STAT3 activation and has reported antiviral and anticancer activity. It is supplied as a high-purity solid with documented solubility and storage profiles for in vitro experimentation and dosing accuracy.

  • Inhibits IL-6-induced STAT3 activation
  • Reported antiviral and anticancer activity
  • High purity (≈99.8%) suitable for research applications
  • Supplied as a white to off-white solid with high solubility in DMSO
  • Stable when stored as powder at -20 °C and in solvent under recommended conditions
  • Molecular weight 730.75 g·mol⁻1 and formula C37H46O15 for accurate dosing

Catalog No. 50-002-64465 Supplier Medchemexpress LLC Supplier No. HY1264215MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Kansuinine A is a natural diterpenoid research compound that inhibits IL-6-induced STAT3 activation and has reported antiviral and anticancer activity. It is supplied as a high-purity solid with documented solubility and storage profiles for in vitro experimentation and dosing accuracy.

  • Inhibits IL-6-induced STAT3 activation
  • Reported antiviral and anticancer activity
  • High purity (≈99.8%) suitable for research applications
  • Supplied as a white to off-white solid with high solubility in DMSO
  • Stable when stored as powder at -20 °C and in solvent under recommended conditions
  • Molecular weight 730.75 g·mol⁻1 and formula C37H46O15 for accurate dosing

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.