Learn More
Enterprise Technology Solutions Kantek Sign Holders,Floor Stand,2 Holders,1-1/2"Dia x 51"H,Steel,BK
SDP
Catalog No. 5000826417
Encompass_Preferred
Floor Stand Sign Holder includes two sign holders to hold 8-1/2" x 11" or 11" x 17" graphics. Heavy-duty construction with steel base and pole is ideal for high-traffic areas. Sign holders feature a snap frame for easy swapping of material. Display panel rotates in portrait or landscape modes and tilts for the best display angle. Mount the display panel at multiple height positions for best visual impact. Floor stand is constructed of coated steel for indoor and outdoor use. Steel pole is 51" high with 1-1/2" diameter. Heavy, 14" diameter steel base provides stability to stand alone or to anchor to the floor (standard hardware included).
Catalog No. 50-008-26417 Supplier Enterprise Technology Solutions Supplier No. KTKSH817
Floor Stand Sign Holder includes two sign holders to hold 8-1/2" x 11" or 11" x 17" graphics. Heavy-duty construction with steel base and pole is ideal for high-traffic areas. Sign holders feature a snap frame for easy swapping of material. Display panel rotates in portrait or landscape modes and tilts for the best display angle. Mount the display panel at multiple height positions for best visual impact. Floor stand is constructed of coated steel for indoor and outdoor use. Steel pole is 51" high with 1-1/2" diameter. Heavy, 14" diameter steel base provides stability to stand alone or to anchor to the floor (standard hardware included).