Learn More
Research Products International Corp Kao & Michayluk Medium, Powder, Makes 1 Liter
SDP
Catalog No. 50213382
Encompass_Preferred
Basal salt mixture containing micro and macro nutrients as described by Kao & Michayluk (1975).
Catalog No. 50-213-382 Supplier Research Products International Corp Supplier No. K201001.0
Basal salt mixture containing micro and macro nutrients as described by Kao & Michayluk (1975).