Learn More
Bioworld Kao and Michayluk Medium, 50 L
Catalog No. 50254634
Encompass_Preferred
Kao & Michayluk MediumContains Macronutrients, Micronutrients, Vitamins, and Organic Acids as described by Kao & Michayluk (1975) Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L or 50L of mediumPlant tissue culture testedWorking Concentration: 3.9 g/LSoluble in water
Catalog No. 50-254-634 Supplier Bioworld Supplier No. 306300403
Kao & Michayluk MediumContains Macronutrients, Micronutrients, Vitamins, and Organic Acids as described by Kao & Michayluk (1975) Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L or 50L of mediumPlant tissue culture testedWorking Concentration: 3.9 g/LSoluble in water