Learn More
Alkali Scientific Kao & Michayluk Medium, Powder, 1 L
SDP
Catalog No. 5000659899
Encompass_Preferred
Kao & Michayluk Medium is a widely used formulation for plant tissue culture, provided in powder form to make 1 liter of solution. This medium supports the growth of a variety of plant species in vitro by supplying necessary nutrients and vitamins that promote cell division and growth. It is ideal for culturing both herbaceous and woody plants, making it a versatile choice in agricultural and botanical research. This medium is commonly used for studying plant regeneration, somatic embryogenesis, and the effects of various growth regulators on plant development. Kao & Michayluk medium is a fundamental tool in plant tissue culture laboratories.
Catalog No. 50-006-59899 Supplier Alkali Scientific Supplier No. KAO001CP
Kao & Michayluk Medium is a widely used formulation for plant tissue culture, provided in powder form to make 1 liter of solution. This medium supports the growth of a variety of plant species in vitro by supplying necessary nutrients and vitamins that promote cell division and growth. It is ideal for culturing both herbaceous and woody plants, making it a versatile choice in agricultural and botanical research. This medium is commonly used for studying plant regeneration, somatic embryogenesis, and the effects of various growth regulators on plant development. Kao & Michayluk medium is a fundamental tool in plant tissue culture laboratories.