Learn More
Haemonetics Corporation Kaolin reagent, blood sample activation, compatible with TEG analyzer, pH 7.4 ± 0.05, contains sodium azide
Supplier:  Haemonetics Corporation 6300
Encompass
Haemonetics Corporation's Kaolin is a specialized reagent aimed at the precise activation of blood samples for coagulation analyses on the TEG analyzer. It acts as a contact activator to standardize the initiation of the intrinsic pathway of hemostasis, ensuring reliable results in diagnostic applications. The formulation includes Kaolin, disodium orthophosphate heptahydrate as a buffer, and sodium azide for preservation, maintaining a controlled pH for stability.
  • Designed for blood sample activation
  • Compatible with TEG analyzer
  • Active ingredient is Kaolin (CAS No 1332-58-7)
  • Buffer system includes disodium orthophosphate heptahydrate (CAS No 7782-85-6)
  • Contains sodium azide as a preservative (CAS No 26628-22-8)
  • Maintains a pH value of 7.4 ± 0.05
  • Not classified as hazardous under normal use
Catalog No. NC0369158