Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kaolin, Monohydrate, Thermo Scientific™

Catalog No. AC412950010
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC412950010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC412950010 Supplier Thermo Scientific Chemicals Supplier No. 412950010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1332-58-7
Molecular Formula Al2H4O9Si2
Molecular Weight (g/mol) 258.16
MDL Number MFCD00062311
InChI Key NLYAJNPCOHFWQQ-UHFFFAOYSA-N
Synonym kaolin, china clay, argilla, glomax, porcelain clay, bolus alba, white bole, hydrated aluminum silicate, kaolin clay slurry, alphagloss
PubChem CID 56841936
IUPAC Name dioxo-2,4,6-trioxa-3,5-disila-1,7-dialuminaheptane-3,5-dione dihydrate
SMILES O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O

Specifications

Color Beige
Physical Form Powder
Quantity 1 kg
Linear Formula H2Al2Si2O8·H2O
Merck Index 14, 5282
Solubility Information 2% max. Soluble Matter (in acid)
Formula Weight 258.16
Grade Extra Pure
Identification positive for aluminum
Loss on Ignition 15% max. (600°C)
Chemical Name or Material Kaolin, monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.