Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kaolin, Powder, USP, Spectrum™ Chemical
SDP

Catalog No. 18603622 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-622 500 g Poly Bottle
18-603-624 2.5 kg Poly Bottle
18-603-623 6 x 500 g Poly Bottle
18-603-625 4 x 2.5 kg Poly Bottle
18-603-626 12 kg Poly Bottle
18-603-627 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-603-622 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. KA105500GM
Add to Cart
Add to Cart

KA105, 1332-58-7

Spectrum™ Chemical Kaolin, Powder, USP is used to treat diarrhea and to relieve soreness and swelling inside the mouth caused by radiation treatments. All Spectrum Chemical manufactured USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1332-58-7
Molecular Formula Al2H4O9Si2
Molecular Weight (g/mol) 258.16
MDL Number MFCD00062311
InChI Key NLYAJNPCOHFWQQ-UHFFFAOYSA-N
IUPAC Name dioxo-2,4,6-trioxa-3,5-disila-1,7-dialuminaheptane-3,5-dione dihydrate
SMILES O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O

Specifications

CAS Max % 100%
Physical Form Crystalline Powder or Needles
Quantity 500 g
Grade USP
Loss on Ignition 15%
Packaging Poly Bottle
Chemical Name or Material Kaolin, Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.