Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Kaolinite

Catalog No. 03584250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-584-250G 250 g
03-584-1KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-584-250G Supplier Sigma Aldrich Supplier No. 03584250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Kaolinite can be used: In the preparation of of polysialate polymers-To prepare mullite composites by sintering kaolinite and alumina.To synthesize microporous materials such as zeolite Linde Type A (LTA) and kaoline based silicoaluminophosphate (SAPO) molecular sieves.In the synthesis of terbium pyridine-picolinate grafted kaoline, as a luminescent hybrid material.As a filler and a coating pigment in the paper industry.

Specifications

CAS 1318-74-7
Molecular Formula Al2O7Si2 -+ 2H2O
Linear Formula Al2O3 -+ 2SiO2 -+ 2H2O
MDL Number MFCD00077645
Quantity 250 g
Synonym Aluminum silicate
Molecular Weight (g/mol) 258.16 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.