Learn More
Bellco Glass, Inc. KAP-UTS/Blue,18mm
SDP
Catalog No. 501945268
Encompass_Preferred
Manufactured from non-toxic autoclavable polypropylene. Ideal for culture tubes and flasks with DeLong style neck and access ports. Same evaporation and sterility characteristics as stainless steel closures. Molded pressure fins for firm grip. There are seven colors and six sizes form 13mm to 38mm. MADE IN THE USA. Case quantity 1000, Color Blue, 18mm
Catalog No. 50-194-5268 Supplier Bellco Glass, Inc. Supplier No. 200718004
Manufactured from non-toxic autoclavable polypropylene. Ideal for culture tubes and flasks with DeLong style neck and access ports. Same evaporation and sterility characteristics as stainless steel closures. Molded pressure fins for firm grip. There are seven colors and six sizes form 13mm to 38mm. MADE IN THE USA. Case quantity 1000, Color Blue, 18mm