Learn More
Roche Diagnostics KAPA 10 mM dNTP Mix, 500 uL
Supplier:  Roche Diagnostics 07958609001
Encompass
Roche offers KAPA premium molecular-grade dNTPs for PCR applications demanding the highest quality dNTPs. KAPA dNTP Mix contains dATP, dCTP, dGTP, and dTTP supplied at a concentration of 10 mm each. *This item is non returnable.
Catalog No. 50-196-5273