Learn More
Roche Diagnostics HotStart ReadyMix with dye | 100x25uL Reactions
Catalog No. 501965212 Shop All Roche Diagnostics Products
Encompass
KAPA2G Fast PCR Kits contain a second-generation (2G) DNA polymerase enzyme engineered for higher processivity and speed, offering significantly faster extension rates than wild-typeTaqpolymerase. In addition to speed, KAPA2G Fast PCR Kit provides higher yields and sensitivity than competitor enzymes across a broad range of targets. *This item is non returnable.
This item is not returnable. View return policy
Catalog No. 50-196-5212 Supplier Roche Diagnostics Supplier No. 07961260001
KAPA2G Fast PCR Kits contain a second-generation (2G) DNA polymerase enzyme engineered for higher processivity and speed, offering significantly faster extension rates than wild-typeTaqpolymerase. In addition to speed, KAPA2G Fast PCR Kit provides higher yields and sensitivity than competitor enzymes across a broad range of targets. *This item is non returnable.