Learn More
Roche Diagnostics KAPA Adapter Dilution Buffer, 25 mL
Supplier:  Roche Diagnostics 08278539001
Encompass
Both KAPA UDI Adapters and KAPA DI Adapters contain standard Illumina adapter backbones and are readily incorporated into sequencing workflows for Illumina sequencers. KAPA UDI Adapters employ 192 non-redundant barcodes for the 96 unique dual-index combinations, whereas KAPA DI Adapters offer a combinatorial approach (twelve i7 x eight i5 barcodes = 96 dual index combinations). *This item is non returnable.
Catalog No. 50-196-5304