Learn More
Roche Diagnostics KAPA EvoPrep Kit DNA Library Preparation Kit, 96 reactions, ReadyMix reagent format, compatible with all Illumina instruments
Catalog No. NC3566295 Shop All Roche Diagnostics Products
Encompass
The KAPA EvoPrep Kit is a high-performance solution for Next-Generation Sequencing (NGS) DNA library preparation, designed to simplify workflows and reduce errors. The kit delivers excellent sequencing performance across various research applications, including whole-genome and targeted sequencing, with enhanced library conversion rates.
  • High-performance DNA library preparation kit
  • Streamlined, automation-friendly workflow
  • Compatible with all Illumina sequencing instruments
  • Validated for a variety of challenging sample inputs
  • High conversion rates up to 100%
  • Flexible input amount ranging from 0.1 ng to 500 ng
  • Storage temperature between -15 °C and -20 °C
  • Includes additional components available separately
  • For research use only, not for diagnostic procedures
Catalog No. NC3566295 Supplier Roche Diagnostics Supplier No. 10096039001
The KAPA EvoPrep Kit is a high-performance solution for Next-Generation Sequencing (NGS) DNA library preparation, designed to simplify workflows and reduce errors. The kit delivers excellent sequencing performance across various research applications, including whole-genome and targeted sequencing, with enhanced library conversion rates.
  • High-performance DNA library preparation kit
  • Streamlined, automation-friendly workflow
  • Compatible with all Illumina sequencing instruments
  • Validated for a variety of challenging sample inputs
  • High conversion rates up to 100%
  • Flexible input amount ranging from 0.1 ng to 500 ng
  • Storage temperature between -15 °C and -20 °C
  • Includes additional components available separately
  • For research use only, not for diagnostic procedures