Learn More
Roche Diagnostics KAPA Mouse Genotyping Kits - Standard kit, 500 reactions
Catalog No. 501965198 Shop All Roche Diagnostics Products
Encompass
KAPA Mouse Gentoyping Kits include KAPA Express Extract Enzyme (1 U/L), KAPA Express Extract Buffer (10X), and KAPA2G Fast (or Fast HotStart) Genotyping Mix (2X), which contains DNA Polymerase, reaction buffer with loading dye, dNTPs, and MgCl2 (at a final concentration of 1.5 mM). ReadyMixes for PCR are also sold separately. *This item is non returnable.
This item is not returnable. View return policy
Catalog No. 50-196-5198 Supplier Roche Diagnostics Supplier No. 07961804001
KAPA Mouse Gentoyping Kits include KAPA Express Extract Enzyme (1 U/L), KAPA Express Extract Buffer (10X), and KAPA2G Fast (or Fast HotStart) Genotyping Mix (2X), which contains DNA Polymerase, reaction buffer with loading dye, dNTPs, and MgCl2 (at a final concentration of 1.5 mM). ReadyMixes for PCR are also sold separately. *This item is non returnable.