Learn More
Roche Diagnostics KAPA mRNA HyperPrep Kit, 24 libraries
Catalog No. 501965251 Shop All Roche Diagnostics Products
Encompass
KAPA RNA HyperPrep Kits include all reagents required for RNA fragmentation, cDNA synthesis, library preparation and amplification. KAPA Pure Beads are supplied in each kit for reaction cleanups, while KAPA Adapters are available separately. Kits also include upfront depletion modules for mRNA capture (KK8580, KK8581), rRNA depletion (KK8560, KK8561), and rRNA and globin depletion (KK8562, KK8563). *This item is non returnable.
This item is not returnable. View return policy
Catalog No. 50-196-5251 Supplier Roche Diagnostics Supplier No. 08098115702
KAPA RNA HyperPrep Kits include all reagents required for RNA fragmentation, cDNA synthesis, library preparation and amplification. KAPA Pure Beads are supplied in each kit for reaction cleanups, while KAPA Adapters are available separately. Kits also include upfront depletion modules for mRNA capture (KK8580, KK8581), rRNA depletion (KK8560, KK8561), and rRNA and globin depletion (KK8562, KK8563). *This item is non returnable.