Learn More
Roche Diagnostics KAPA mRNA HyperPrep (96rxn) Kit
Catalog No. 501965308 Shop All Roche Diagnostics Products
Encompass
KAPA mRNA HyperPrep Kits include all reagents required for mRNA Capture, RNA fragmentation, cDNA synthesis, library preparation and amplification. KAPA Pure Beads are supplied in each kit for reaction cleanups while KAPA Adapters are available separately. KAPA mRNA HyperPrep (96rxn) Kit includes three component kits: mRNA Capture 96 rxn Kit, RNA Hyper 96 rxn Kit, and 3 x 5mL Pure Beads. *This item is non returnable.
This item is not returnable. View return policy
Catalog No. 50-196-5308 Supplier Roche Diagnostics Supplier No. 08098123702
KAPA mRNA HyperPrep Kits include all reagents required for mRNA Capture, RNA fragmentation, cDNA synthesis, library preparation and amplification. KAPA Pure Beads are supplied in each kit for reaction cleanups while KAPA Adapters are available separately. KAPA mRNA HyperPrep (96rxn) Kit includes three component kits: mRNA Capture 96 rxn Kit, RNA Hyper 96 rxn Kit, and 3 x 5mL Pure Beads. *This item is non returnable.