Learn More
Roche Diagnostics KAPA RNA HyperPrep Kit, 24 libraries
Catalog No. 501965276 Shop All Roche Diagnostics Products
Encompass
KAPA RNA HyperPrep Kits include all reagents required for RNA fragmentation, cDNA synthesis, library preparation and amplification. KAPA Pure Beads are supplied in each kit for reaction cleanups, while KAPA Adapters are available separately. *This item is non returnable.
This item is not returnable. View return policy
Catalog No. 50-196-5276 Supplier Roche Diagnostics Supplier No. 08098093702
KAPA RNA HyperPrep Kits include all reagents required for RNA fragmentation, cDNA synthesis, library preparation and amplification. KAPA Pure Beads are supplied in each kit for reaction cleanups, while KAPA Adapters are available separately. *This item is non returnable.