Learn More
Roche Diagnostics KAPA RNA HyperPrep Kit with RiboErase (HMR) Globin, 96 Libraries, RNA-Seq, 25 ng - 1 μg input RNA, for high- and low-quality RNA, including FFPE tissue
Catalog No. NC3448457 Shop All Roche Diagnostics Products
Encompass
The KAPA RNA HyperPrep Kit with RiboErase (HMR) Globin is a streamlined, strand-specific solution designed for high-quality RNA-seq library preparation from various RNA inputs. This kit reduces preparation time and combines enzymatic steps while ensuring comprehensive transcriptome coverage by efficiently depleting ribosomal RNA and globin mRNA from samples. It is suitable for both manual and automated workflows on Illumina platforms and is compatible with degraded samples, making it an ideal choice for various research applications.
  • Streamlined library preparation reduces time
  • Strand-specific RNA-seq libraries
  • Compatible with low-input and degraded samples
  • Supports both manual and automated workflows
  • Efficient rRNA and globin mRNA depletion
  • High throughput and consistent results
  • Designed for Illumina platforms
  • Kit accommodates 96 libraries
  • Research use only
Catalog No. NC3448457 Supplier Roche Diagnostics Supplier No. 08308241702
Add to Cart
Add to Cart
Request a Quote
The KAPA RNA HyperPrep Kit with RiboErase (HMR) Globin is a streamlined, strand-specific solution designed for high-quality RNA-seq library preparation from various RNA inputs. This kit reduces preparation time and combines enzymatic steps while ensuring comprehensive transcriptome coverage by efficiently depleting ribosomal RNA and globin mRNA from samples. It is suitable for both manual and automated workflows on Illumina platforms and is compatible with degraded samples, making it an ideal choice for various research applications.
  • Streamlined library preparation reduces time
  • Strand-specific RNA-seq libraries
  • Compatible with low-input and degraded samples
  • Supports both manual and automated workflows
  • Efficient rRNA and globin mRNA depletion
  • High throughput and consistent results
  • Designed for Illumina platforms
  • Kit accommodates 96 libraries
  • Research use only