Learn More
Roche Diagnostics KAPA SYBR FAST qPCR Kits - 1000 x 20 uL reactions, ROX (ABI Prism), 10 mL
Catalog No. 501965199 Shop All Roche Diagnostics Products
Encompass
Kits include KAPA SYBR FAST qPCR Master Mix (2X), which contains KAPA SYBR FAST DNA Polymerase, reaction buffer, dNTPs, SYBR Green I dye, and MgCl2 at a final concentration of 2.5 mM. *This item is non returnable.
This item is not returnable. View return policy
Catalog No. 50-196-5199 Supplier Roche Diagnostics Supplier No. 07959435001
Kits include KAPA SYBR FAST qPCR Master Mix (2X), which contains KAPA SYBR FAST DNA Polymerase, reaction buffer, dNTPs, SYBR Green I dye, and MgCl2 at a final concentration of 2.5 mM. *This item is non returnable.