Learn More
Roche Diagnostics KAPA SYBR FAST qPCR Kits - 500 x 20 uL reactions, ROX (ABI Prism), 5 mL
Catalog No. 501965208 Shop All Roche Diagnostics Products
Encompass
Kits include KAPA SYBR FAST qPCR Master Mix (2X), which contains KAPA SYBR FAST DNA Polymerase, reaction buffer, dNTPs, SYBR Green I dye, and MgCl2 at a final concentration of 2.5 mM. *This item is non returnable.
This item is not returnable. View return policy
Catalog No. 50-196-5208 Supplier Roche Diagnostics Supplier No. 07959427001
Kits include KAPA SYBR FAST qPCR Master Mix (2X), which contains KAPA SYBR FAST DNA Polymerase, reaction buffer, dNTPs, SYBR Green I dye, and MgCl2 at a final concentration of 2.5 mM. *This item is non returnable.