Learn More
Roche Diagnostics KAPA Taq PCR Kit | 250U
Supplier:  Roche Diagnostics 07958765001
Encompass
The Kapa Taq DNA polymerase enzyme is based on the single-subunit, wild-typeTaqDNA polymerase of the thermophilic bacteriumThermus aquaticus. Kapa Taq and Kapa Taq HotStart DNA polymerase enzymes have 5-3 polymerase and 5-3 exonuclease activities, but no 3 to 5 exonuclease (proofreading) activity. The enzyme has an error rate of approximately 1 error per 2.2 x 105nucleotides incorporated. *This item is non returnable.
Catalog No. 50-196-5238