Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappa Goat F(ab')2 anti-Mouse, LE/AF (Low Endotoxin/Azide Free), Polyclonal, Southern Biotech™
SDP

Catalog No. OB105214 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB105214 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB105214 Supplier Southern Biotechnology Associates Supplier No. 105214
Only null left
Add to Cart
Add to Cart

Goat F(ab')2 Polyclonal Secondary Antibody

SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.

Specifications

Antigen Kappa
Applications ELISA, Dot Blot, Flow Cytometry, Functional Assay
Classification Low Endotoxin/Azide-free (LE/AF)
Concentration 0.5mg/mL
Formulation Supplied in PBS
Host Species Goat
Immunogen Mouse κ light chains
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at -20°C
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.