Learn More
Diagnostic Biosystems KAPPA LIGHT CHAIN Kappa Light Chain
Catalog No. 502407424
Encompass_Preferred
This antibody reacts with free as well as bound kappa light chains. Contaminating antibodies have been removed by solid phase adsorption.
Catalog No. 50-240-7424 Supplier Diagnostic Biosystems Supplier No. PDR015
This antibody reacts with free as well as bound kappa light chains. Contaminating antibodies have been removed by solid phase adsorption.