Learn More
Kappa Light Chain Antibody (RM103), Biotin, Novus Biologicals™

Description
Specifications
Specifications
| Antigen | Kappa Light Chain |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | RM103 |
| Concentration | 1 mg/mL |
| Conjugate | Biotin |
| Dilution | Western Blot 0.1ug/ml - 0.5ug/ml, Flow Cytometry, ELISA 0.005ug/ml-0.2ug/ml, Immunohistochemistry 0.5 - 2 ug/mL, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunocytochemistry 0.5 - 2 ug/mL |
| Formulation | PBS, 50% Glycerol, 1% BSA with 0.09% Sodium Azide |
| Gene Alias | HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 |
| Gene Symbols | IGKC |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.