Learn More
Diagnostic Biosystems KAPPA LIGHT CHAIN-FITC Kappa Light Chain-FITC
Supplier:  Diagnostic Biosystems F001
Encompass_Preferred
Traces of contaminating antibodies have been removed by solid phase absorption. Only the kappa precipitation line appeared in cross immunoelectrophoresis. This antibody is well suited for tissue section staining but may be used for other applications.
Catalog No. 50-240-7345