Learn More
Sigma Aldrich Fine Chemicals Biosciences Kappa Light Chains free fr1MG
Catalog No. 502904334
Encompass
kappa Light Chains (free) is obtained from human IgG kappa myeloma sera. fractionation ion-exchange and affinity chromatography procedures are used for purification.
Catalog No. 50-290-4334 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. K33881MG
kappa Light Chains (free) is obtained from human IgG kappa myeloma sera. fractionation ion-exchange and affinity chromatography procedures are used for purification.