Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappler™ Frontline 300 Chemical and Fire Protection Bib Trousers
SDP

Catalog No. 18025681 Shop All Kappler, Inc. Products
Change view
Click to view available options
Size:
2X-Large/3X-Large
Large/X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-025-681 2X-Large/3X-Large
18-025-680 Large/X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-025-681 Supplier Kappler™ Supplier No. F3H660 SI 2X3X
This item is not returnable. View return policy

Specifications

Closure Type Adjustable Webbing Straps with Snap-lock
Color Silver
Material Frontline 300
Seam Type Heat-sealed, Taped
Size 2X-Large/3X-Large
For Use With (Application) Petrochemical operations, line maintenance, tank cleaning, refueling situations
Certifications/Compliance ASTM F1001
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.