Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappler™ Frontline 300 Chemical and Fire Protection Jacket
SDP

Catalog No. 18025678 Shop All Kappler, Inc. Products
Change view
Click to view available options
Closure Type:
Double Hook and Loop Storm Flap, Zipper
Hook and Loop Storm Flap, Zipper
Size:
2X-Large/3X-Large
Large/X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type Size
18-025-678 Hook and Loop Storm Flap, Zipper Large/X-Large
18-025-679 Double Hook and Loop Storm Flap, Zipper 2X-Large/3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-025-678 Supplier Kappler™ Supplier No. F3H675 SI LGXL
This item is not returnable. View return policy

Specifications

Size Large/X-Large
Material Frontline 300
Color Silver
Closure Type Hook and Loop Storm Flap, Zipper
Construction Heat Sealed and Taped Seam
Cuff Style Cone inserts
Seam Type Sealed
For Use With (Application) Petrochemical operations, line maintenance, tank cleaning, refueling situations
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.