Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappler™ Frontline 300 Flat Back Half C Chemical and Fire Protection Jacket and Hood
SDP

Catalog No. 18025685 Shop All Kappler, Inc. Products
Change view
Click to view available options
Size:
2X-Large/3X-Large
Large/X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-025-685 2X-Large/3X-Large
18-025-684 Large/X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-025-685 Supplier Kappler™ Supplier No. F3H761 SI 2X3X

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Specifications

Size 2X-Large/3X-Large
Material CPE, PVC
Color Silver
Closure Type Double Hook and Loop Storm Flap, Zipper
Certifications/Compliance ASTM F1001
Construction Heat Sealed and Taped Seam, PVC Visor, CPE Cuffs
Cuff Style CPE
Seam Type Sealed
Size (English) Inseam 30 in.
Size (Metric) Inseam 76.2 cm
Thickness 1.01 mm
For Use With (Application) Petrochemical operations, line maintenance, tank cleaning, refueling situations
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.