Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappler™ Zytron™ 100 Chemical-Resistant Coveralls
SDP

Catalog No. 19167835C Shop All Kappler, Inc. Products
Change view
Click to view available options
Serged Seams; Collar, open wrists and ankles; Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-167-835C Serged Seams; Collar, open wrists and ankles; Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-167-835C Supplier Kappler™ Supplier No. Z1S412L

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides basic chemical resistance

  • Zippered front
  • Seam type available in bound and serged
  • Available with attached hood and/or boots
  • Color: Yellow

Ideal for applications where workers need protection from dry particulates and light splash

Compliance

Meets ASTM™ F739 industrial chemical testing requirements.

Specifications

Size Large
Material Zytron™
Color Yellow
Ankle Style Open
Cuff Style Open
Closure Type Front Zipper
Seam Type Serged
Resistance Chemicals
Certifications/Compliance ASTM D751, D3787
Description Collar, open wrists and ankles
For Use With (Application) Ideal for applications where workers need protection from dry particulates and light splash
Weight (English) 12 lb.
Weight (Metric) 5.4 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.