Learn More
Cole-Parmer Film, 2 Mil, no adhesive, 12'' x 12''
Supplier:  Cole-Parmer 0827786
Encompass_Preferred
Kapton Type HN film is a tough, aromatic polyimide film, exhibiting an excellent balance of physical, chemical, and electrical properties over a wide temperature range, particularly at unusually high temperatures. It has been used successfully in applications at temperatures as low as -452°F (-269°C) and as high as 752°F (400°C). HN film can be laminated, metallized, punched, formed or adhesive coated.
Catalog No. NC0511796