Learn More
Premier Lab Supply Kapton 7.5μm (0.30mil) Sheets – PN# TF-475-345
SDP
Catalog No. NC0846029
Encompass_Preferred
Kapton film 7.5μm (0.30mil) Sheets with Dimensions: 3” x 3” (76 x 76mm) (PN# TF-475-345). Kapton has excellent chemical resistance and moderate transmission characteristics. Pure polymide film, is virtually free of trace contaminants. Lot number tracking system. 500 per box.
Catalog No. NC0846029 Supplier Premier Lab Supply Supplier No. TF475345
Kapton film 7.5μm (0.30mil) Sheets with Dimensions: 3” x 3” (76 x 76mm) (PN# TF-475-345). Kapton has excellent chemical resistance and moderate transmission characteristics. Pure polymide film, is virtually free of trace contaminants. Lot number tracking system. 500 per box.