Learn More
Cole-Parmer Polymide Tape, 1" W; 36 yd/RL, 1 RL/Pk
Supplier:  Cole-Parmer 0827782
Encompass_Preferred
Polymide tape is a chemical-resistant tape that is manufactured with a high temperature silicone adhesive. Provides high heat resistance and tensile strength, good solvent resistance, and excellent dielectric strength. Temperature range is -100 to 500°F (-73 to 260°C). Meets UL-510 for flame retardant specifications.
Catalog No. NC9540770