Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uline KAPTON TAPE 1 MIL, 2"" X 36 YDS

Catalog No. NC0912752
Encompass

Kapton Tape - 1 Mil, 2 in. x 36 yds

Catalog No. NC0912752 Supplier Uline Supplier No. S11731
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Kapton Tape - 1 Mil, 2 in. x 36 yds

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.