Learn More
Uline Kapton® Tape - 1 Mil, 1 1⁄4" x 36 yds
Catalog No. NC2855215
Encompass
Maximum resistance to high temperatures.Recommended for masking circuit boards and splicing wires.Temperature range up to 500°F.Polyimide film with silicone adhesive for easy release.UL 510 flame retardant.
Catalog No. NC2855215 Supplier Uline Supplier No. S17210
Maximum resistance to high temperatures.Recommended for masking circuit boards and splicing wires.Temperature range up to 500°F.Polyimide film with silicone adhesive for easy release.UL 510 flame retardant.