Learn More
McMaster-Carr Masking Tape for Electronics
Catalog No. NC2855350
Encompass
Masking Tape for Electronics Silicone, Kapton Polyimide, 3/4" Wide,.0025" Thickness, 108' Long
Catalog No. NC2855350 Supplier McMaster-Carr Supplier No. 7648A33
Masking Tape for Electronics Silicone, Kapton Polyimide, 3/4" Wide,.0025" Thickness, 108' Long