Learn More
Electron Microscopy Sciences Kapton Thin Polyimide Tape, 1/2" wide
SDP
Catalog No. NC1181414
Encompass_Preferred
Kapton Polyimide Tape is known for its superior ability to maintain its physical properties under harsh conditions. This general purpose film easily bonds to difficult surfaces with silicone adhesive, and leaves little to no residue when removed. Microscopists find it ideal for holding samples or masking during deposition in vacuum systems (it is relatively low outgassing). It is also perfect for positioning samples in cryo or high temperature working conditions. There are no known organic solvents for the film. Tape thickness with thinner layer of adhesive: 2.5 mil. No liner.
Width: ½"
Catalog No. NC1181414 Supplier Electron Microscopy Sciences Supplier No. 7770807
Kapton Polyimide Tape is known for its superior ability to maintain its physical properties under harsh conditions. This general purpose film easily bonds to difficult surfaces with silicone adhesive, and leaves little to no residue when removed. Microscopists find it ideal for holding samples or masking during deposition in vacuum systems (it is relatively low outgassing). It is also perfect for positioning samples in cryo or high temperature working conditions. There are no known organic solvents for the film. Tape thickness with thinner layer of adhesive: 2.5 mil. No liner.
Width: ½"