Learn More
Medchemexpress LLC Karanjin | 521-88-0 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN25345MG
Encompass_Preferred
Karanjin is an orally active furanoflavonoid isolated from Leguminosae plants. It exhibits a range of biological activities including anti-diabetic, anti-cancer, anti-inflammatory, antioxidant, and neuroprotective properties. It also functions as an insect repellent and acaricide, making it suitable for various research applications.
  • Orally active furanoflavonoid
  • Isolated from Leguminosae
  • Anti-diabetic, anti-cancer, anti-inflammatory, antioxidant properties
  • Neuroprotective, anti-ulcer, and anticolitis effects
  • Insect repellent, insecticidal, and acaricide activity
  • Enhances glucose uptake; inhibits PTPase
  • Induces apoptosis; blocks cell cycle in cancer cells
  • Decreases reactive oxygen species (ROS)
  • Reduces blood glucose and joint injury in animal models
  • Improves learning and memory
Catalog No. 50-004-19734