Learn More
Medchemexpress LLC Karanjin | 521-88-0 | MFCD00017406 | 99.9% | 292.29 g/mol | C18H12O4 | 10 MG
SDP
Catalog No. 5000289595
Encompass_Preferred
Karanjin is a natural furanoflavonol used in research to modulate cellular pathways and induce apoptosis. It is reported to have antidiabetic, anti-inflammatory, antioxidant, and anticancer activities and is supplied for in vitro pharmacology and pathway modulation studies.
  • Natural furanoflavonol with bioactive properties.
  • Modulates AMPK and can induce apoptosis in cell-based assays.
  • Reported activities include antidiabetic, anti-inflammatory, antioxidant, and anticancer effects.
  • High purity suitable for research use; soluble in DMSO.
  • Available in small mass pack sizes tailored for laboratory experiments.
Catalog No. 50-002-89595 Supplier Medchemexpress LLC Supplier No. HYN253410MG
Add to Cart
Add to Cart
Karanjin is a natural furanoflavonol used in research to modulate cellular pathways and induce apoptosis. It is reported to have antidiabetic, anti-inflammatory, antioxidant, and anticancer activities and is supplied for in vitro pharmacology and pathway modulation studies.
  • Natural furanoflavonol with bioactive properties.
  • Modulates AMPK and can induce apoptosis in cell-based assays.
  • Reported activities include antidiabetic, anti-inflammatory, antioxidant, and anticancer effects.
  • High purity suitable for research use; soluble in DMSO.
  • Available in small mass pack sizes tailored for laboratory experiments.