Learn More
Essendant KARAT FOOD BUCKET W/LID 170OZ
Catalog No. NC2939253
Encompass_Preferred
Karat Food Bucket Bundles are great for storing family sized meals. Included matching lids are flat paper lids. The lids fit in the groove on top of the bucket which makes it perfect for storage and being on-the-go. Made with PE lining which makes it great for greasy food to go such as popcorn, fried chicken and even pastries! Food Container Type: Bucket/Lid Combo; Material(s): Paper; PE Lining; Material Type: Paper; Interior Volume Capacity: 170 oz.
Catalog No. NC2939253 Supplier Essendant Supplier No. KRTCFB170WBNDL
Karat Food Bucket Bundles are great for storing family sized meals. Included matching lids are flat paper lids. The lids fit in the groove on top of the bucket which makes it perfect for storage and being on-the-go. Made with PE lining which makes it great for greasy food to go such as popcorn, fried chicken and even pastries! Food Container Type: Bucket/Lid Combo; Material(s): Paper; PE Lining; Material Type: Paper; Interior Volume Capacity: 170 oz.