Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SW Sustainability Solutions KarbonHex™ KX70 Professional Built Cut-Resistant Winter Gloves
SDP

Catalog No. 17441144 Shop All SW Sustainability Solutions Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-441-144 Medium
17-441-145 Large
17-441-146 X-Large
17-441-147 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 17-441-144 Supplier SW Sustainability Solutions Supplier No. C244653
Add to Cart
Add to Cart
This item is not returnable. View return policy

Durable, heavyweight, winter-lined, seamless handling gloves offer warmth and dryness with a textured foam latex knuckle dipped coating for secure wet and dry grip.

  • Heavyweight seamless 10 g knitted thermal shell with brushed fleece lining provides all-day warmth and comfort.
  • Sandy foam latex 3/4 coating provides excellent wet and dry grip while protecting from water and liquids.
  • Elastic knit wrist ensures a comfortable and secure fit.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI-ISEA 105-16
Certifications/Compliance (Europe) EN388:2016
Coated Area 3/4 dip
Construction Seamless Knit
Cuff Material Acrylic, Elastic
Coating Material Latex
Color Black, Gray
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free No
Material Latex, Polyester
Product Type Mechanical Protection Gloves
Size Medium
EN388 Standard EN388:3X41B
For Use With (Application) Mechanical Applications
Gender Unisex
Glove Cuff Color Black
Glove Cuff Style Elasticated
Grip Design Sandpatch
Liner Material Acrylic/Polyester
Lining Seamless Lining
Model C244653
Puncture Rating ANSI/ISEA 105-16 Level 2
Series C24465
Thumb Style Straight
Yarn Gauge 10 G
Length (English) 10.2 in.
Length (Metric) 25.9 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.