Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SW Sustainability Solutions KarbonHex™ KX84V Professional Built Cut-Resistant Winter Gloves, Hi-Vis Cold Protection
SDP

Catalog No. 17441151 Shop All SW Sustainability Solutions Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-441-151 2X-Large
17-441-148 Medium
17-441-149 Large
17-441-150 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 17-441-151 Supplier SW Sustainability Solutions Supplier No. C334616
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Seamless cut-resistant, durable, and heavyweight hi-vis winter lined gloves offer warmth and dryness. Textured foam latex with 75% dipped coating for secure wet and dry grip.

  • Cut-resistant, hi-vis, heavyweight, seamless 10 g knitted thermal shell with brushed fleece lining for all day warmth and comfort.
  • Sandy foam latex 3/4 coating provides excellent wet and dry grip while protecting from water and liquids.
  • Elastic knit wrist ensures a comfortable and secure fit.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI-ISEA 105-16
Certifications/Compliance (Europe) EN388:2016/ EN511
Coated Area 3/4 dip
Construction Seamless Knit
Coating Material Latex
Color Hi-vis Yellow
Cut Resistance Rating ANSI A3
Hand Specific
Latex Free No
Material Latex, Nylon, Glass Fiber
Product Type Cut-resistant Gloves
Size 2X-Large
EN388 Standard EN388:1X31C
For Use With (Application) Mechanical Applications
Gender Unisex
Glove Cuff Style Elasticated
Grip Design Sandpatch
Liner Material Acrylic/Glass Fiber/Nylon
Lining Seamless Lining
Model C334616
Puncture Rating ANSI/ISEA 105-16 Level 2
Series C33461
Thumb Style Straight
Yarn Gauge 10 G
Length (English) 10.2 in.
Length (Metric) 25.9 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.