Learn More
Medchemexpress LLC HY-14812 5mg Medchemexpress, Karenitecin CAS:203923-89-1 Purity:>98%
SDP
Catalog No. 502081645
Encompass_Preferred
Medchemexpress, HY-14812 5mg Karenitecin CAS:203923-89-1 Karenitecin (Cositecan) is a topoisomerase I inhibitor, with potent anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1645 Supplier Medchemexpress LLC Supplier No. HY148125MG
Medchemexpress, HY-14812 5mg Karenitecin CAS:203923-89-1 Karenitecin (Cositecan) is a topoisomerase I inhibitor, with potent anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.