Learn More
Medchemexpress LLC Kari 201 hydrochloride | 2376132-24-8 | 320.86 | 5 MG
SDP
Catalog No. 5000386466
Encompass_Preferred
KARI 201 hydrochloride is a selective, brain penetrant pan-competitive acid sphingomyelinase (ASM) inhibitor with an IC50 of 338.3 nM. It also acts as a ghrelin receptor agonist.
  • Selective, brain penetrant pan-competitive acid sphingomyelinase (ASM) inhibitor
  • Ghrelin receptor agonist
  • Improves neuropathological features of Alzheimer's disease
Catalog No. 50-003-86466 Supplier Medchemexpress LLC Supplier No. HY171962A5MG
KARI 201 hydrochloride is a selective, brain penetrant pan-competitive acid sphingomyelinase (ASM) inhibitor with an IC50 of 338.3 nM. It also acts as a ghrelin receptor agonist.
  • Selective, brain penetrant pan-competitive acid sphingomyelinase (ASM) inhibitor
  • Ghrelin receptor agonist
  • Improves neuropathological features of Alzheimer's disease