Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Karl Fischer Aqualine™ Electrolyte AG, for Karl Fischer Titration By Coulometry, Fisher Chemical™

Catalog No. AL2520500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AL-2520500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AL-2520500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nonhalogenated anolyte solution for general use in conventional cells with a diaphragm

  • Anolyte solution for use in conventional cells with a diaphragm
  • Source of electrolyte required for anodic oxidation for coulometric analysis
  • Contains methanol as a solvent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aqualine™ Electrolyte AG
Name Note Karl Fischer Reagent
Color Colorless
CAS 111-42-2, 7553-56-2, 108-75-8, 7446-09-5
Physical Form Liquid
Quantity 500 mL
Packaging Amber glass bottle
MDL Number 3097387
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.