Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Karl Fischer Coulometric CG, catholyte solution, Thermo Scientific™
Click to view available options
Quantity:
6 x 110 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Specifications
Color | Colorless to Yellow |
Chemical Name or Material | Karl Fischer Coulometric CG |
Quantity | 6 x 110 mL |
Flash Point | 92°C (198°F) |
Sensitivity | Moisture sensitive |
Density | 1.1 g/mL |
Name Note | Catholyte solution |
Boiling Point | 202°C |
Physical Form | Liquid |
UN Number | UN3267 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction