Learn More
GFS Chemicals Inc WATERMARK KARL FISCHER REAGENT, PYRIDINE-FREE SINGLE SOLUTION, 5 mg/ml | 1 L | GFS CHEMICALS, INC. Item #: 99601 | Bulk #: 1600
SDP
Catalog No. NC9698569
Encompass_Preferred
WATERMARK KARL FISCHER REAGENT, PYRIDINE-FREE SINGLE SOLUTION, 5 mg/ml | For volumetric analysis. Suitable for general purpose or aldehydes and ketones. ; Specifications : Suitability, Titer Strength (when bottled) (mg/ml) 4.7 - 5.5 mg/ml
This item is not returnable. View return policy
Catalog No. NC9698569 Supplier GFS Chemicals Inc Supplier No. 99601
WATERMARK KARL FISCHER REAGENT, PYRIDINE-FREE SINGLE SOLUTION, 5 mg/ml | For volumetric analysis. Suitable for general purpose or aldehydes and ketones. ; Specifications : Suitability, Titer Strength (when bottled) (mg/ml) 4.7 - 5.5 mg/ml