Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Karl Fischer Reagent, for pyridine-based volumetric one-component KF titration, Solstice

Catalog No. 60046637 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-637 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 60-046-637 Supplier Solstice Supplier No. 361151LUS

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Solutions are prepared with raw materials which meet or exceed requirements of the American Chemical Society
  • Where no ACS specifications exist, chemicals of highest purity available are used
  • All aqueous solutions are prepared using high purity, deionized, water which meets requirements for ASTM type 1 Reagent Grade Water
  • Exacting production and quality control records are maintained providing traceability of raw materials
  • All solutions are guaranteed to meet label specifications
  • All solution labels meet or exceed all ISO, DOT and OSHA regulations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.10
MDL Number MFCD00081734
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
IUPAC Name 2-methoxyethan-1-ol
SMILES COCCO

Specifications

Boiling Point 123°C
CAS Min % ≥10.00, ≥20.00
CAS Max % <20.00, <10.00
CAS 7553-56-2, 110-86-1, 109-86-4, 7446-09-5
Density 1.150 g/cm3
Physical Form Liquid
Quantity 1 L
Flash Point 33°C
Packaging Polycoated Bottle
Grade One Component Volumetry
Hazard Category Danger
Hazard Statement H226+H302+H312+H332+H314+H351+H360+H370+H372
Precautionary Statement P241+P242+P243+P260+P264+P270+P271+P280+P240+P233+P210+P202+P201
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.